MilliporeSigma
  • Seasonal expressions of prostaglandin E synthases and receptors in the prostate of the wild ground squirrel (Spermophilus dauricus).
Seasonal expressions of prostaglandin E synthases and receptors in the prostate of the wild ground squirrel (Spermophilus dauricus).
Prostaglandins & other lipid mediators (2020-01-14)
Zhu Chen, Yuchen Yao, Yuyao Shen, Ao Zhang, Yue Zhang, Wenqian Xie, Yu Wang, Yingying Han, Zhengrong Yuan, Qiang Weng, Haolin Zhang
ABSTRACT
The prostate gland is a male accessory reproductive gland, whose vitality and function are under tight regulation of different hormones. Prostaglandins E2 (PGE2) is one of the major products generated by the actions of cyclooxygenases (COX) and prostaglandin E synthase (PTGES) on arachidonic acid, and is involved in a number of physiological and pathological processes. In this study, we investigated the seasonal immunolocalizations and expressions of COX-1, COX-2 and PTGES, as well as PGE2 receptors (PTGERs) subtypes 1-4 (EP1, EP2, EP3, EP4) in the prostate of the wild ground squirrel. Histological examination observed enlarged prostatic lumens in the breeding season and significantly shrunken lumens in the nonbreeding season. COX-1, COX-2, PTGES and PTGERs were mainly localized in epithelial and stromal cells in the breeding and nonbreeding seasons. The mRNA expression levels of Cox-1, Cox-2, Ptges, Ptger2 (encoding EP2) and Ptger4 (encoding EP4) were higher in the prostate of the breeding season than in the nonbreeding season. The relative mRNA levels of Cox-1, Cox-2, Ptges, Ptger2 and Ptger4 were positively correlated with prostatic weights. In addition, both the prostatic and plasma concentrations of PGE2 were significantly higher in the breeding season compared to the nonbreeding season. These results suggested that PGE2 synthesis and signaling might play an important autocrine or paracrine role in the regulation of seasonal changes in the prostatic function of the wild ground squirrel.