MilliporeSigma
  • Infrared driven hot electron generation and transfer from non-noble metal plasmonic nanocrystals.
Infrared driven hot electron generation and transfer from non-noble metal plasmonic nanocrystals.
Nature communications (2020-06-12)
Dongming Zhou, Xufeng Li, Qiaohui Zhou, Haiming Zhu
ABSTRACT
Non-noble metal plasmonic materials, e.g. doped semiconductor nanocrystals, compared to their noble metal counterparts, have shown unique advantages, including broadly tunable plasmon frequency (from visible to infrared) and rich surface chemistry. However, the fate and harvesting of hot electrons from these non-noble metal plasmons have been much less explored. Here we report plasmon driven hot electron generation and transfer from plasmonic metal oxide nanocrystals to surface adsorbed molecules by ultrafast transient absorption spectroscopy. We show unambiguously that under infrared light excitation, hot electron transfers in ultrafast timescale (<50 fs) with an efficiency of 1.4%. The excitation wavelength and fluence dependent study indicates that hot electron transfers right after Landau damping before electron thermalization. We revealed the efficiency-limiting factors and provided improvement strategies. This study paves the way for designing efficient infrared light absorption and photochemical conversion applications based on non-noble metal plasmonic materials.
Sigma-Aldrich
Cadmium acetylacetonate, ≥99.9% trace metals basis