MilliporeSigma
  • Cas12a Base Editors Induce Efficient and Specific Editing with Low DNA Damage Response.
Cas12a Base Editors Induce Efficient and Specific Editing with Low DNA Damage Response.
Cell reports (2020-06-04)
Xiao Wang, Chengfeng Ding, Wenxia Yu, Ying Wang, Siting He, Bei Yang, Yi-Chun Xiong, Jia Wei, Jifang Li, Jiayi Liang, Zongyang Lu, Wei Zhu, Jing Wu, Zhi Zhou, Xingxu Huang, Zhen Liu, Li Yang, Jia Chen
ABSTRACT
The advent of base editors (BEs) holds great potential for correcting pathogenic-related point mutations to treat relevant diseases. However, Cas9 nickase (nCas9)-derived BEs lead to DNA double-strand breaks, which can trigger unwanted DNA damage response (DDR). Here, we show that the original version of catalytically dead Cas12a (dCas12a)-conjugated BEs induce a basal level of DNA breaks and minimally activate DDR proteins, including H2AX, ATM, ATR, and p53. By fusing dCas12a with engineered human apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A (APOBEC3A), we further develop the BEACON (base editing induced by human APOBEC3A and Cas12a without DNA break) system to achieve enhanced deamination efficiency and editing specificity. Efficient C-to-T editing is achieved by BEACON in mammalian cells at levels comparable to AncBE4max, with only low levels of DDR and minimal RNA off-target mutations. Importantly, BEACON induces in vivo base editing in mouse embryos, and targeted C-to-T conversions are detected in F0 mice.
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)