MilliporeSigma
  • Asymmetric Disulfanylbenzamides as Irreversible and Selective Inhibitors of Staphylococcus aureus Sortase A.
Asymmetric Disulfanylbenzamides as Irreversible and Selective Inhibitors of Staphylococcus aureus Sortase A.
ChemMedChem (2020-03-03)
Fabian Barthels, Gabriella Marincola, Tessa Marciniak, Matthias Konhäuser, Stefan Hammerschmidt, Jan Bierlmeier, Ute Distler, Peter R Wich, Stefan Tenzer, Dirk Schwarzer, Wilma Ziebuhr, Tanja Schirmeister
ABSTRACT
Staphylococcus aureus is one of the most frequent causes of nosocomial and community-acquired infections, with drug-resistant strains being responsible for tens of thousands of deaths per year. S. aureus sortase A inhibitors are designed to interfere with virulence determinants. We have identified disulfanylbenzamides as a new class of potent inhibitors against sortase A that act by covalent modification of the active-site cysteine. A broad series of derivatives were synthesized to derive structure-activity relationships (SAR). In vitro and in silico methods allowed the experimentally observed binding affinities and selectivities to be rationalized. The most active compounds were found to have single-digit micromolar Ki values and caused up to a 66 % reduction of S. aureus fibrinogen attachment at an effective inhibitor concentration of 10 μM. This new molecule class exhibited minimal cytotoxicity, low bacterial growth inhibition and impaired sortase-mediated adherence of S. aureus cells.
Sigma-Aldrich
Gly-Gly-Gly-Gly