MilliporeSigma
  • Saliva exosomes-derived UBE2O mRNA promotes angiogenesis in cutaneous wounds by targeting SMAD6.
Saliva exosomes-derived UBE2O mRNA promotes angiogenesis in cutaneous wounds by targeting SMAD6.
Journal of nanobiotechnology (2020-05-08)
Bobin Mi, Lang Chen, Yuan Xiong, Chenchen Yan, Hang Xue, Adriana C Panayi, Jing Liu, Liangcong Hu, Yiqiang Hu, Faqi Cao, Yun Sun, Wu Zhou, Guohui Liu
ABSTRACT
Enhancing angiogenesis is critical for accelerating wound healing. Application of different types of exosomes (Exos) to promote angiogenesis represents a novel strategy for enhanced wound repair. Saliva is known to accelerate wound healing, but the underlying mechanisms remain unclear. Our results have demonstrated that saliva-derived exosomes (saliva-Exos) induce human umbilical vein endothelial cells (HUVEC) proliferation, migration, and angiogenesis in vitro, and promote cutaneous wound healing in vivo. Further experiments documented that Ubiquitin-conjugating enzyme E2O (UBE2O) is one of the main mRNAs of saliva-Exos, and activation of UBE2O has effects similar to those of saliva-Exos, both in vitro and in vivo. Mechanistically, UBE2O decreases the level of SMAD family member 6 (SMAD6), thereby activating bone morphogenetic protein 2 (BMP2), which, in turn, induces angiogenesis. The present work suggests that administration of saliva-Exos and UBE2O represents a promising strategy for enhancing wound healing through promotion of angiogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human UBE2O
Pricing and availability is not currently available.