MilliporeSigma
  • Upregulation of OSBPL3 by HIF1A promotes colorectal cancer progression through activation of RAS signaling pathway.
Upregulation of OSBPL3 by HIF1A promotes colorectal cancer progression through activation of RAS signaling pathway.
Cell death & disease (2020-07-28)
Hong-Li Jiao, Bin-Shu Weng, Shan-Shan Yan, Zi-Mo Lin, Shu-Yang Wang, Xiao-Ping Chen, Guang-Hua Liang, Xiao-Qing Li, Wei-Yi Zhao, Jia-Yi Huang, Dan Zhang, Ling-Jie Zhang, Fang-Yi Han, Sheng-Nan Li, Li-Jie Chen, Jiong-Hua Zhu, Wen-Feng He, Yan-Qing Ding, Ya-Ping Ye
ABSTRACT
Oxysterol-binding protein like protein 3 (OSBPL3) has been shown involving in the development of several human cancers. However, the relationship between OSBPL3 and colorectal cancer (CRC), particularly the role of OSBPL3 in the proliferation, invasion and metastasis of CRC remains unclear. In this study, we investigated the role of OSBPL3 in CRC and found that its expression was significantly higher in CRC tissues than that in normal tissues. In addition, high expression of OSBPL3 was closely related to poor differentiation, advanced TNM stage and poor prognosis of CRC. Further experiments showed that over-expression of OSBPL3 promoted the proliferation, invasion and metastasis of CRC in vitro and in vivo models. Moreover, we revealed that OSBPL3 promoted CRC progression through activation of RAS signaling pathway. Furthermore, we demonstrated that hypoxia induced factor 1 (HIF-1A) can regulate the expression of OSBPL3 via binding to the hypoxia response element (HRE) in the promoter of OSBPL3. In summary, Upregulation of OSBPL3 by HIF1A promotes colorectal cancer progression through activation of RAS signaling pathway. This novel mechanism provides a comprehensive understanding of both OSBPL3 and the RAS signaling pathway in the progression of CRC and indicates that the HIF1A-OSBPL3-RAS axis is a potential target for early therapeutic intervention in CRC progression.
Sigma-Aldrich
GGTI-2133, ≥98% (HPLC), solid