MilliporeSigma
  • Implication of HYBID (Hyaluronan-Binding Protein Involved in Hyaluronan Depolymerization) in Hyaluronan Degradation by Synovial Fibroblasts in Patients with Knee Osteoarthritis.
Implication of HYBID (Hyaluronan-Binding Protein Involved in Hyaluronan Depolymerization) in Hyaluronan Degradation by Synovial Fibroblasts in Patients with Knee Osteoarthritis.
The American journal of pathology (2020-02-23)
Jun Shiozawa, Susana de Vega, Mehmet Z Cilek, Chiho Yoshinaga, Tomomi Nakamura, Shinya Kasamatsu, Hiroyuki Yoshida, Haruka Kaneko, Muneaki Ishijima, Kazuo Kaneko, Yasunori Okada
ABSTRACT
Cell migration-inducing hyaluronidase 1 (CEMIP), also known as hyaluronan (HA)-binding protein involved in HA depolymerization (HYBID), plays a role in HA degradation. CEMIP2, also known as transmembrane protein 2 (TMEM2), possessing a sequence similarity with HYBID, is reported as a hyaluronidase in mice. However, the expression of these molecules in osteoarthritic synovium and their involvement in HA degradation in synovial fluid (SF) from patients with knee osteoarthritis remain elusive. This study examined their expression in synovial tissue and the relationship with molecular weight of HA in SF in knee osteoarthritis patients. Quantification of mRNA demonstrated that HYBID expression is significantly (5.5-fold) higher in osteoarthritic synovium than in normal control synovium, whereas TMEM2 expression level is similar between the two groups. By immunohistochemistry, HYBID was localized mainly to CD68-negative and fibroblast-specific protein 1-positive synovial lining cells and sublining fibroblasts in osteoarthritic synovium. The mRNA expression levels of HYBID, but not TMEM2, in osteoarthritic synovium positively correlated with distribution of lower-molecular-weight HA with below 1000 kDa in SF. HA-degrading activity in osteoarthritic synovial fibroblasts was abrogated by siRNA-mediated knockdown of HYBID. Among the 12 factors examined, IL-6 significantly up-regulated the HYBID expression and HA-degrading activity in osteoarthritic synovial fibroblasts. These data suggest that HYBID overexpressed by IL-6-stimulated synovial fibroblasts is implicated in HA degradation in osteoarthritic synovium.
Sigma-Aldrich
Anti-KIAA1199 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.