MilliporeSigma
  • Bismuth Vanadate/Bilirubin Oxidase Photo(bio)electrochemical Cells for Unbiased, Light-Triggered Electrical Power Generation.
Bismuth Vanadate/Bilirubin Oxidase Photo(bio)electrochemical Cells for Unbiased, Light-Triggered Electrical Power Generation.
ChemSusChem (2020-02-19)
Dina Mukha, Yifat Cohen, Omer Yehezkeli
ABSTRACT
The construction of bias- and donor-free photobioelectrochemical cells for the generation of light-triggered electrical power is presented. The developed oxygen reduction biocathodes are based on bilirubin oxidase (BOD) that originates from Myrothecium verrucaria (MvBOD) and a thermophilic Bacillus pumilus (BpBOD). Methods to entrap the BOD with 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) redox molecules in a polydopamine layer are presented. A pH-independent, positively charged pyrenebetaine linker was synthesized, utilized, and led to a threefold improvement to the bioelectrocatalytic current. Both the developed polydopamine/ABTS/MvBOD and the pyrenebetaine/BpBOD biocathodes were further coupled with BiVO4 /cobalt phosphate water-oxidation photoanodes to construct biotic/abiotic photobioelectrochemical cells, which generated power outputs of 0.74 and 0.85 mW cm-2 , respectively. The presented methods are versatile, show the strength of biotic/abiotic hybrids, and can be further used to couple different redox enzymes with electrodes.
Sigma-Aldrich
1-Aminopyrene, 97%
Pricing and availability is not currently available.