MilliporeSigma
  • Novel talc encapsulated lanthanum alginate hydrogel for efficient phosphate adsorption and fixation.
Novel talc encapsulated lanthanum alginate hydrogel for efficient phosphate adsorption and fixation.
Chemosphere (2020-05-26)
Bin Wang, Wei Zhang, Lu Li, Wenbin Guo, Jie Xing, Hongyu Wang, Xiaoling Hu, Wanlin Lyu, Rongfan Chen, Jianyang Song, Ling Chen, Zhenzhen Hong
ABSTRACT
In the present work, talc (a low-cost clay) encapsulated salts alginate (TAL) beads were synthesized by cross-linking with lanthanum ion and tested for phosphate adsorption. Multiple methods were applied for the characterization of composites. The combined effect of talc and lanthanum improved phosphate removal performance of TAL beads. Factors such as talc content, La3+ concentration, adsorbent dosage, pH, co-existing ions (Cl-, NO3- and SO42-) were studied in batch experiments. The optimized TAL-7 beads exhibited satisfactory selectivity towards phosphate in the coexistence of competing anions and could remain efficient phosphate removal in the pH range of 4-6. The phosphate removal efficiency reached to 95% with a maximum uptake of 16.4 mg P/g obtained at the optimal pH 4. Further experiments suggested that Langmuir isotherm model and the pseudo-second-order kinetic model could well describe the phosphate adsorption process of TAL-7 beads. Moreover, TAL-7 beads exhibited superior phosphate fixation performance in the long-term experiment. The results from adsorption experiment and characterization analysis demonstrated that TAL-7 beads could be a cost-effective and promising biosorbent for phosphate adsorption and fixation in the aqueous environment.
Sigma-Aldrich
Lanthanum(III) chloride hydrate, 99.9% trace metals basis