MilliporeSigma
  • The association between microRNA-21 and hypertension-induced cardiac remodeling.
The association between microRNA-21 and hypertension-induced cardiac remodeling.
PloS one (2020-02-11)
Ken Watanabe, Taro Narumi, Tetsu Watanabe, Yoichiro Otaki, Tetsuya Takahashi, Tomonori Aono, Jun Goto, Taku Toshima, Takayuki Sugai, Masahiro Wanezaki, Daisuke Kutsuzawa, Shigehiko Kato, Harutoshi Tamura, Satoshi Nishiyama, Hiroki Takahashi, Takanori Arimoto, Tetsuro Shishido, Masafumi Watanabe
ABSTRACT
Hypertension is a major public health problem among the aging population worldwide. It causes cardiac remodeling, including hypertrophy and interstitial fibrosis, which leads to development of hypertensive heart disease (HHD). Although microRNA-21 (miR-21) is associated with fibrogenesis in multiple organs, its contribution to cardiac remodeling in hypertension is poorly understood. Circulating miR-21 level was higher in patients with HHD than that in the control subjects. It also positively correlated with serum myocardial fibrotic markers. MiR-21 expression levels were significantly upregulated in the mice hearts after angiotensin II (Ang II) infusion or transverse aortic constriction (TAC) compared with control mice. Expression level of programmed cell death 4 (PDCD4), a main target of miR-21, was significantly decreased in Ang II infused mice and TAC mice compared with control mice. Expression levels of transcriptional activator protein 1 (AP-1) and transforming growth factor-β1 (TGF-β1), which were downstream targets of PDCD4, were increased in Ang II infused mice and TAC mice compared with control mice. In vitro, mirVana-miR-21-specific inhibitor attenuated Ang II-induced PDCD4 downregulation and contributed to subsequent deactivation of AP-1/TGF-β1 signaling pathway in neonatal rat cardiomyocytes. Thus, suppression of miR-21 prevents hypertrophic stimulation-induced cardiac remodeling by regulating PDCD4, AP-1, and TGF-β1 signaling pathway.
Sigma-Aldrich
MISSION® esiRNA, targeting human PDCD4