MilliporeSigma
Signalling through the platelet glycoprotein Ib-V-IX complex.
Cellular signalling (2004-09-24)
Ilaria Canobbio, Cesare Balduini, Mauro Torti
ABSTRACT
The glycoprotein Ib-V-IX is one of the major adhesive receptors expressed on the surface of circulating platelets. It is composed of four different polypeptides-GPIbalpha, GPIbbeta, GPIX, and GPV-and represents a multifunctional receptor able to interact with a number of ligands, including the adhesive protein von Willebrand factor, the coagulation factors thrombin, factors XI and XII, and the membrane glycoproteins P-selectin and Mac-1. Interaction of GPIb-V-IX with the subendothelial von Willebrand factor is essential for primary haemostasis, as it initiates platelet adhesion to the subendothelial matrix at the sites of vascular injury even under high flow conditions. Upon interaction with von Willebrand factor, GPIb-V-IX initiates transmembrane signalling events for platelet activation, which eventually result in integrin alpha(IIb)beta(3) stimulation and platelet aggregation. The investigation of the biochemical mechanisms for platelet activation by GPIb-V-IX has attracted increasing attention during the last years. This review will describe and discuss recent findings that have provided new insights into the events underlying GPIb-V-IX transmembrane signalling. In particular, it will summarise basic concepts on the structure of this receptor, extracellular ligands, and intracellular interactors potentially involved in transmembrane signalling. The recently suggested role of membrane Fc receptors in GPIb-V-IX-initiated platelet activation will also be discussed, along with the involvement of lipid metabolising enzymes, tyrosine kinases, and the cytoskeleton in the crosstalk between GPIb-V-IX and integrin alpha(IIb)beta(3).