MilliporeSigma
  • Determination of ibuprofen and flurbiprofen in pharmaceuticals by capillary zone electrophoresis.
Determination of ibuprofen and flurbiprofen in pharmaceuticals by capillary zone electrophoresis.
Journal of pharmaceutical and biomedical analysis (2006-05-12)
Rafifa Hamoudová, Marie Pospísilová
ABSTRACT
Capillary zone electrophoresis with spectrophotometric detection was used for the determination of ibuprofen (IB) and flurbiprofen (FL) in pharmaceuticals. The separation was carried out in a fused silica capillary (60 cm x 100 microm i.d. effective length 45 cm) at 30 kV with UV detection at 232 nm. The optimized background electrolyte was 20mM N-(2-acetamido)-2-aminoethanesulfonic acid (ACES) with 20mM imidazole and 10mM alpha-cyclodextrin of pH 7.3. 2-Naphthoxyacetic acid was used as internal standard. A single analysis took less than 5 min. Rectilinear calibration ranges were 2-500 mg l(-1) for IB and 1-60 mg l(-1) for FL. The relative standard deviations (R.S.D.) values (n=6) were 1.53% for IB and 1.29% for FL (for 200 mg l(-1) IB and 10 mg l(-1) FL). This validated method has been successfully applied for the routine analysis of 10 commercially available pharmaceutical preparations (syrup, tablets, cream and gel).
Sigma-Aldrich
ACES, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.