MilliporeSigma
  • PIWIL4 regulates cervical cancer cell line growth and is involved in down-regulating the expression of p14ARF and p53.
PIWIL4 regulates cervical cancer cell line growth and is involved in down-regulating the expression of p14ARF and p53.
FEBS letters (2012-04-10)
Chang Su, Zhong-Jie Ren, Fang Wang, Min Liu, Xin Li, Hua Tang
ABSTRACT
Recently, PIWIL4 has been identified as a functional protein involved in tumorigenesis. Cervical cancer is the second most prevalent form of cancer worldwide. The relationship between PIWIL4 and cervical cancer is still unknown. Here, we found that PIWIL4 is up-regulated in human cervical cancer tissues in comparison to adjacent normal tissues, and it promotes cell growth and proliferation by inhibiting apoptosis through the p14ARF/p53 pathway. PIWIL4 can also promote the invasion of cervical cancer cells. These results suggest that PIWIL4 might play an oncogenic role in cervical cancer and be useful as a new therapeutic target in the future.