MilliporeSigma
  • Dissecting ubiquitin signaling with linkage-defined and protease resistant ubiquitin chains.
Dissecting ubiquitin signaling with linkage-defined and protease resistant ubiquitin chains.
Angewandte Chemie (International ed. in English) (2014-09-10)
Tatjana Schneider, Daniel Schneider, Daniel Rösner, Saurav Malhotra, Franziska Mortensen, Thomas U Mayer, Martin Scheffner, Andreas Marx
ABSTRACT
Ubiquitylation is a complex posttranslational protein modification and deregulation of this pathway has been associated with different human disorders. Ubiquitylation comes in different flavors: Besides mono-ubiquitylation, ubiquitin chains of various topologies are formed on substrate proteins. The fate of ubiquitylated proteins is determined by the linkage-type of the attached ubiquitin chains, however, the underlying mechanism is poorly characterized. Herein, we describe a new method based on codon expansion and click-chemistry-based polymerization to generate linkage-defined ubiquitin chains that are resistant to ubiquitin-specific proteases and adopt native-like functions. The potential of these artificial chains for analyzing ubiquitin signaling is demonstrated by linkage-specific effects on cell-cycle progression.