MilliporeSigma
  • Simultaneous catabolite repression between glucose and toluene metabolism in Pseudomonas putida is channeled through different signaling pathways.
Simultaneous catabolite repression between glucose and toluene metabolism in Pseudomonas putida is channeled through different signaling pathways.
Journal of bacteriology (2007-07-10)
Teresa del Castillo, Juan L Ramos
ABSTRACT
Pseudomonas putida KT2440(pWW0) can use toluene via the TOL plasmid-encoded catabolic pathways and can use glucose via a series of three peripheral chromosome-encoded routes that convert glucose into 6-phosphogluconate (6PG), namely, the glucokinase pathway, in which glucose is transformed to 6PG through the action of glucokinase and glucose-6-phosphate dehydrogenase. Alternatively, glucose can be oxidized to gluconate, which can be phosphorylated by gluconokinase to 6PG or oxidized to 2-ketogluconate, which, in turn, is converted into 6PG. Our results show that KT2440 metabolizes glucose and toluene simultaneously, as revealed by net flux analysis of [(13)C]glucose. Determination of glucokinase and gluconokinase activities in glucose metabolism, gene expression assays using a fusion of the promoter of the Pu TOL upper pathway to 'lacZ, and global transcriptomic assays revealed simultaneous catabolite repression in the use of these two carbon sources. The effect of toluene on glucose metabolism was directed to the glucokinase branch and did not affect gluconate metabolism. Catabolite repression of the glucokinase pathway and the TOL pathway was triggered by two different catabolite repression systems. Expression from Pu was repressed mainly via PtsN in response to high levels of 2-dehydro-3-deoxygluconate-6-phosphate, whereas repression of the glucokinase pathway was channeled through Crc.
Sigma-Aldrich
3-Deoxy-2-keto-6-phosphogluconic acid lithium salt, ≥95% (TLC)
Pricing and availability is not currently available.