MilliporeSigma
Solution-Phase Synthesis of Cesium Lead Halide Perovskite Nanowires.
Journal of the American Chemical Society (2015-07-17)
Dandan Zhang, Samuel W Eaton, Yi Yu, Letian Dou, Peidong Yang
ABSTRACT
Halide perovskites have attracted much attention over the past 5 years as a promising class of materials for optoelectronic applications. However, compared to hybrid organic-inorganic perovskites, the study of their pure inorganic counterparts, like cesium lead halides (CsPbX3), lags far behind. Here, a catalyst-free, solution-phase synthesis of CsPbX3 nanowires (NWs) is reported. These NWs are single-crystalline, with uniform growth direction, and crystallize in the orthorhombic phase. Both CsPbBr3 and CsPbI3 are photoluminescence active, with composition-dependent temperature and self-trapping behavior. These NWs with a well-defined morphology could serve as an ideal platform for the investigation of fundamental properties and the development of future applications in nanoscale optoelectronic devices based on all-inorganic perovskites.
Sigma-Aldrich
Cesium bromide, AnhydroBeads, 99.999% trace metals basis, (Perovskite grade)