MilliporeSigma
  • Pharmacokinetics of BAY 59-7939--an oral, direct Factor Xa inhibitor--in rats and dogs.
Pharmacokinetics of BAY 59-7939--an oral, direct Factor Xa inhibitor--in rats and dogs.
Xenobiotica; the fate of foreign compounds in biological systems (2005-11-26)
C Weinz, U Buetehorn, H-P Daehler, C Kohlsdorfer, U Pleiss, S Sandmann, K-H Schlemmer, T Schwarz, W Steinke
ABSTRACT
The pharmacokinetics of BAY 59-7939 - a novel, oral, direct Factor Xa inhibitor - were investigated in rats and dogs in support of preclinical safety studies and clinical development. BAY 59-7939 was rapidly absorbed after oral dosing, with an absolute bioavailability of 57-66% in rats, and 60-86% in dogs. Plasma pharmacokinetics of BAY 59-7939 were linear across the investigated dose range (1-10 mg kg(-1) in rats, 0.3-3 mg kg(-1) in dogs). Plasma clearance was low: 0.4 l kg(-1) h(-1) in rats and 0.3 l kg(-1) h(-1) in dogs; volume of distribution (V(ss)) was moderate: 0.3 l kg(-1) in rats, and 0.4 l kg(-1) in dogs. The elimination half-life after oral administration was short in both species (0.9-2.3 h). Whole-body autoradiography showed moderate tissue affinity. No retention or small volume enrichments of BAY 59-7939-related radioactivity were observed. The plasma-protein binding of BAY 59-7939 was high, species dependent and fully reversible. BAY 59-7939 was rapidly excreted in rats and dogs, and was not irreversibly retained. A dual mode of excretion (biliary/faecal and renal) was observed. In summary, BAY 59-7939 had a favourable, predictable pharmacokinetic profile, with high oral bioavailability and a dual route of excretion.
Sigma-Aldrich
Rivaroxaban, ≥98% (HPLC)