MilliporeSigma
  • Asynchronous mixing of kidney progenitor cells potentiates nephrogenesis in organoids.
Asynchronous mixing of kidney progenitor cells potentiates nephrogenesis in organoids.
Communications biology (2020-05-13)
Ashwani Kumar Gupta, Prasenjit Sarkar, Jason A Wertheim, Xinchao Pan, Thomas J Carroll, Leif Oxburgh
ABSTRACT
A fundamental challenge in emulating kidney tissue formation through directed differentiation of human pluripotent stem cells is that kidney development is iterative, and to reproduce the asynchronous mix of differentiation states found in the fetal kidney we combined cells differentiated at different times in the same organoid. Asynchronous mixing promoted nephrogenesis, and heterochronic organoids were well vascularized when engrafted under the kidney capsule. Micro-CT and injection of a circulating vascular marker demonstrated that engrafted kidney tissue was connected to the systemic circulation by 2 weeks after engraftment. Proximal tubule glucose uptake was confirmed, but despite these promising measures of graft function, overgrowth of stromal cells prevented long-term study. We propose that this is a technical feature of the engraftment procedure rather than a specific shortcoming of the directed differentiation because kidney organoids derived from primary cells and whole embryonic kidneys develop similar stromal overgrowth when engrafted under the kidney capsule.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Human Nuclei Antibody, clone 235-1, Biotin Conjugate, clone 235-1, from mouse, biotin conjugate
Pricing and availability is not currently available.