MilliporeSigma
  • Renal functional effects of the highly selective AT2R agonist, β-Pro7 Ang III, in normotensive rats.
Renal functional effects of the highly selective AT2R agonist, β-Pro7 Ang III, in normotensive rats.
Clinical science (London, England : 1979) (2020-03-24)
Lucinda M Hilliard Krause, Brandon A Kemp, Amanda Suan Jui Tan, Emma S Jones, Mark P Del Borgo, Marie-Isabel Aguilar, Kate M Denton, Robert M Carey, Robert E Widdop
ABSTRACT
Recently, we designed a group of peptides by sequential substitution of the naturally occurring α-amino acid throughout the Ang III peptide sequence with the corresponding β-amino acid. β-Amino acid substitution at the proline residue of Ang III (β-Pro7-Ang III) resulted in a highly selective AT2R ligand, demonstrating remarkable selectivity for the AT2R in both binding and functional studies. To provide additional functional evidence for the suitability of β-Pro7 Ang III as a novel AT2R agonist, we tested effects of acute systemic administration of β-Pro7-Ang III on renal hemodynamic and excretory function in anesthetized normotensive male and female rats. We also compared the natriuretic effects of acute intrarenal administration of native Ang III and β-Pro7-Ang III in the presence of systemic AT1R blockade in anesthetized female rats to allow for the differentiation of systemic versus direct intrarenal natriuretic actions of β-Pro7-Ang III. In both male and female rats, acute systemic administration of β-Pro7-Ang III elicited renal vasodilatation and natriuresis. Notably, greater renal vasodilatory effects were observed in female versus male rats at the highest dose of β-Pro7-Ang III administered. Moreover, intra-renal administration of β-Pro7-Ang III produced significant natriuretic effects in female rats and, like Ang III, evoked AT2R translocation to the apical plasma membrane in renal proximal tubular cells. Taken together, our findings support the use of β-Pro7-Ang III as a novel AT2R agonist and experimental tool for exploring AT2R function and its potential as a therapeutic target. Furthermore, our findings provide further evidence of a sex-specific influence of AT2R stimulation on renal function.
Sigma-Aldrich
Anti-AT2 Receptor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Nerve Growth Factor Receptor Antibody, p75, serum, Chemicon®