MilliporeSigma
  • Surface functionalization using catalyst-free azide-alkyne cycloaddition.
Surface functionalization using catalyst-free azide-alkyne cycloaddition.
Bioconjugate chemistry (2010-10-23)
Alexander Kuzmin, Andrei Poloukhtine, Margreet A Wolfert, Vladimir V Popik
ABSTRACT
The utility of catalyst-free azide-alkyne [3 + 2] cycloaddition for the immobilization of a variety of molecules onto a solid surface and microbeads was demonstrated. In this process, the surfaces are derivatized with aza-dibenzocyclooctyne (ADIBO) for the immobilization of azide-tagged substrates via a copper-free click reaction. Alternatively, ADIBO-conjugated molecules are anchored to the azide-derivatized surface. Both immobilization techniques work well in aqueous solutions and show excellent kinetics under ambient conditions. We report an efficient synthesis of aza-dibenzocyclooctyne (ADIBO), thus far the most reactive cyclooctyne in cycloaddition to azides. We also describe convenient methods for the conjugation of ADIBO with a variety of molecules directly or via a PEG linker.
Sigma-Aldrich
dPEG®48-biotin acid, >90% (HPLC)
Sigma-Aldrich
dPEG®24-biotin acid, >95% (HPLC)
Sigma-Aldrich
dPEG®4-biotin acid, >95% (HPLC)