MilliporeSigma
  • Comparative efficacy and safety of chlorthalidone and hydrochlorothiazide-meta-analysis.
Comparative efficacy and safety of chlorthalidone and hydrochlorothiazide-meta-analysis.
Journal of human hypertension (2019-10-09)
Stela Dineva, Katya Uzunova, Velichka Pavlova, Elena Filipova, Krassimir Kalinov, Toni Vekov
ABSTRACT
Hypertension is a complex syndrome of multiple hemodynamic, neuroendocrine, and metabolic abnormalities. The goals of treatment in hypertension are to optimally control high blood pressure and to reduce associated cardiovascular morbidity and mortality using the most suitable therapy available. Hydrochlorothiazide (HCTZ) and chlorthalidone (CTLD) are with proven hypertensive effects. The topic of our meta-analysis is to compare the efficacy of HCTZ and CTLD therapy in patient with hypertension. A search of electronic databases PubMed, MEDLINE, Scopus, PsyInfo, eLIBRARY.ru was performed. We chose the random-effects method for the analysis and depicted the results as forest plots. Sensitivity analyses were performed in order to evaluate the degree of significance of each study. Of the 1289 identified sources, only nine trials directly compared HCTZ and CTLD and were included in the meta-analysis. Changes in SBP lead to WMD (95% CI) equal to -3.26 mmHg showing a slight but statistically significant prevalence of CTLD. Results from analyzed studies referring to DBP lead to WMD (95% CI) equal to -2.41 mmHg, which is also statistically significant. During our analysis, we found that there were not enough studies presenting enough data on the effect of CTLD and HCTZ on levels of serum potassium and serum sodium. Our meta-analysis has demonstrated a slight superiority for CTLD regarding blood pressure control. At the same time, the two medications do not show significant differences in their safety profile.
Sigma-Aldrich
Chlorothiazide, thiazide diuretic
Pricing and availability is not currently available.