MilliporeSigma
  • Determination of enantiomeric impurity of linezolid by capillary electrophoresis using heptakis-(2,3-diacetyl-6-sulfato)-beta-cyclodextrin.
Determination of enantiomeric impurity of linezolid by capillary electrophoresis using heptakis-(2,3-diacetyl-6-sulfato)-beta-cyclodextrin.
Journal of chromatography. A (2008-01-01)
Katarzyna Michalska, Genowefa Pajchel, Stefan Tyski
ABSTRACT
A method for the enantioseparation of linezolid, the first compound of a truly new class of antibiotics-the oxazolidinones, was developed. The elaborated method of linezolid enantiomers separation was successfully performed using an anionic single-isomer cyclodextrin-heptakis-(2,3-diacetyl-6-sulfato)-beta-cyclodextrin (HDAS-beta-CD) as a resolving agent with the help of the charged resolving agent migration model (CHARM model). The best results were obtained with 27.5mM HDAS-beta-CD dissolved in 50mM borate buffer, pH 9.0, 15 degrees C, normal polarity. The facile strategies for the reversal of the enantiomers elution order are also described. Afterwards, the optimized method was validated in terms of sensitivity, linearity, accuracy and precision.
Sigma-Aldrich
β-Cyclodextrin hydrate, 99%