MilliporeSigma
  • Immune efficacy of carbon nanotubes recombinant subunit vaccine against largemouth bass ulcerative syndrome virus.
Immune efficacy of carbon nanotubes recombinant subunit vaccine against largemouth bass ulcerative syndrome virus.
Fish & shellfish immunology (2020-03-17)
Yi-Jun Jia, Zi-Rao Guo, Rui Ma, De-Kui Qiu, Zhao Zhao, Gao-Xue Wang, Bin Zhu
ABSTRACT
Largemouth bass ulcerative syndrome virus (LBUSV) is an important virus induce the mortality of largemouth bass (Micropterus Salmoides). In this study, we reported a single-walled carbon nanotubes (SWCNTs) containing LBUSV major capsid protein (MCP) subunit vaccine (SWCNTs-MCP) which was evaluated for its protective effect on largemouth bass by immersion immunization. We found an elevation in serum antibody levels, enzyme activities, complement C3 content and immune-related genes (IgM, TGF-β, IL-1β, IL-8, TNF-α and CD4) expression, in the SWCNTs-MCP immunized groups compared with the pure MCP group. The survival rates for control group, pure MCP protein groups (40 mg L-1) and four SWCNTs-MCP groups (5 mg L-1, 10 mg L-1, 20 mg L-1 and 40 mg L-1) were 0%, 27.78%, 30.56%, 50.00%, 66.67% and 80.56%, respectively. The results suggests that with the assistance of SWCNTs, the immune protection of the SWCNTs-MCP group (40 mg L-1) increased up 52.78%-80.1% compared with pure MCP group (40 mg L-1). Our results demonstrate that the single-walled carbon nanotube subunit vaccine can be used as a new immunization method against LBUSV showing protection following challenge with LBUSV. Taken together, our results demonstrate that the single-walled carbon nanotube subunit vaccine can be used as a new method against LBUSV and have a high immune protection during the largemouth bass farm.