MilliporeSigma
  • Structural relationships for the design of responsive azobenzene-based lyotropic liquid crystals.
Structural relationships for the design of responsive azobenzene-based lyotropic liquid crystals.
Physical chemistry chemical physics : PCCP (2020-02-08)
Luke W Giles, Joshua B Marlow, Calum S G Butler, Geosmin A Turpin, Liliana de Campo, Stephen T Mudie, Charl F J Faul, Rico F Tabor
ABSTRACT
Light-responsive binary (azobenzene + solvent) lyotropic liquid crystals (LCs) were investigated by structural modification of simple azobenzene molecules. Three benzoic acid-containing azobenzene molecules 4-(4-(hydroxyphenyl)diazenyl)benzoic acid (AZO1), 3-(4-(hydroxyphenyl)diazenyl)benzoic acid (AZO2) and 5-(4-(hydroxyphenyl)diazenyl)isophthalic acid (AZO3) were produced with various amide substitutions to produce tectons with a variety of hydrophobicity, size and branching. The LC mesophases formed by binary (azobenzene + solvent) systems with low volatility solvents dimethylsulfoxide (DMSO) and N,N-dimethylformamide (DMF) as well as the protic ionic liquids ethylammonium formate (EAF) and propylammonium formate (PAF), were investigated using a combination of small-angle X-ray and neutron scattering (SAXS and SANS) as well as polarising light microscopy (PLM). Increasing alkyl group length was found to have a strong influence on LC phase spacing, and changes in the position of substitution on the benzene ring influenced the preferred curvature of phases. UV-induced trans to cis isomerization of the samples was shown to influence ordering and optical birefringence, indicating potential applications in optical devices.
Sigma-Aldrich
4-Aminotetrahydropyran, 97%