MilliporeSigma
  • Multi-target optimization of solid phase microextraction to analyse key flavour compounds in wort and beer.
Multi-target optimization of solid phase microextraction to analyse key flavour compounds in wort and beer.
Food chemistry (2020-03-03)
Ana C Vieira, Ana C Pereira, José C Marques, Marco S Reis
ABSTRACT
Despite the literature comprises numerous studies dealing with the analysis of wort and beer flavour-related compounds by HS-SPME followed by GC-MS quantification, no generalized consensus exists regarding the optimal conditions for the extraction procedure. The complex chemistry nature of these matrices, the number of analytes, as well as the number and interactions among parameters affecting the extraction performance, requires the adoption of optimal experimental design protocols. This aspect is often overlooked and often not properly addressed in practice. Therefore, in the present work, the optimal conditions under which a range of wort and beer analytes can be extracted and quantified were analysed. The optimal extraction conditions were presented at two levels of aggregation: global (untargeted) and key-flavour analysis. Experimental data was generated by Definitive-Screening-Design, followed by model development and optimization. Both approaches were compared and critically analysed. For vicinal-diketones group, a complete validation study for the optimal conditions is presented.
Sigma-Aldrich
4-Methyl-1-pentanol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Pricing and availability is not currently available.