MilliporeSigma
  • Differential Effect of Long-Term Systemic Exposure of TNFα on Health of the Annulus Fibrosus and Nucleus Pulposus of the Intervertebral Disc.
Differential Effect of Long-Term Systemic Exposure of TNFα on Health of the Annulus Fibrosus and Nucleus Pulposus of the Intervertebral Disc.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research (2019-12-05)
Deborah J Gorth, Olivia K Ottone, Irving M Shapiro, Makarand V Risbud
ABSTRACT
The inflammatory cytokine tumor necrosis factor alpha (TNFα) is considered to play a key role in the pathogenesis of intervertebral disc disease. To evaluate the importance of this cytokine we examined the inflammatory environment and spinal phenotype of 9-month-old human TNFα overexpressing transgenic (hTNFα-TG) mice. The mice evidenced increased circulating levels of interleukin-1β (IL-1β), IL-2, keratinocyte chemoattractant/human growth-regulated oncogene (KC/GRO), and monocyte chemoattractant protein-1 (MCP-1) along with thinning of the cortical and trabecular vertebral bone. Surprisingly, although the nucleus pulposus (NP) of these mice was intact and healthy, the caudal annulus fibrosus (AF) evidenced robust cell death and immune cell infiltration. Despite these differences, there were no obvious alterations in the collagen or aggrecan content in the NP and AF. However, there was a reduction in cartilage oligomeric matrix protein (COMP), suggesting destabilization of the AF matrix. Microarray analysis of the NP from hTNFα-TG mice cells revealed minimal changes in global gene expression. These findings lend support to the notion that NP tissue is isolated from systemic inflammation. In contrast, the severe AF phenotype suggests that systemic inflammation interferes with AF health, predisposing discs to herniation as opposed to directly causing NP degeneration. © 2020 American Society for Bone and Mineral Research.
Sigma-Aldrich
Pellet pestles, Cordless motor
Sigma-Aldrich
Anti-Aggrecan Antibody, Chemicon®, from rabbit