MilliporeSigma
  • Nanostructured front electrodes for perovskite/c-Si tandem photovoltaics.
Nanostructured front electrodes for perovskite/c-Si tandem photovoltaics.
Optics express (2020-04-01)
Ihteaz M Hossain, Yidenekachew J Donie, Raphael Schmager, Mohamed S Abdelkhalik, Michael Rienäcker, Tobias F Wietler, Robby Peibst, Andrei Karabanov, Jonas A Schwenzer, Somayeh Moghadamzadeh, Ulrich Lemmer, Bryce S Richards, Guillaume Gomard, Ulrich W Paetzold
ABSTRACT
The rise in the power conversion efficiency (PCE) of perovskite solar cells has triggered enormous interest in perovskite-based tandem photovoltaics. One key challenge is to achieve high transmission of low energy photons into the bottom cell. Here, nanostructured front electrodes for 4-terminal perovskite/crystalline-silicon (perovskite/c-Si) tandem solar cells are developed by conformal deposition of indium tin oxide (ITO) on self-assembled polystyrene nanopillars. The nanostructured ITO is optimized for reduced reflection and increased transmission with a tradeoff in increased sheet resistance. In the optimum case, the nanostructured ITO electrodes enhance the transmittance by ∼7% (relative) compared to planar references. Perovskite/c-Si tandem devices with nanostructured ITO exhibit enhanced short-circuit current density (2.9 mA/cm2 absolute) and PCE (1.7% absolute) in the bottom c-Si solar cell compared to the reference. The improved light in-coupling is more pronounced for elevated angle of incidence. Energy yield enhancement up to ∼10% (relative) is achieved for perovskite/c-Si tandem architecture with the nanostructured ITO electrodes. It is also shown that these nanostructured ITO electrodes are also compatible with various other perovskite-based tandem architectures and bear the potential to improve the PCE up to 27.0%.
Sigma-Aldrich
Formamidinium iodide