MilliporeSigma
  • Methods for the reliable induction of heterotopic ossification in the conditional Alk2Q207D mouse.
Methods for the reliable induction of heterotopic ossification in the conditional Alk2Q207D mouse.
Journal of musculoskeletal & neuronal interactions (2020-03-07)
Haichun Pan, Nicole Fleming, Charles C Hong, Yuji Mishina, Daniel S Perrien
ABSTRACT
Conditional Alk2Q207D-floxed (caALK2fl) mice have previously been used as a model of heterotopic ossification (HO). However, HO formation in this model can be highly variable, and it is unclear which methods reliably induce HO. Hence, these studies report validated methods for reproducibly inducing HO in caALK2fl mice. Varying doses of Adex-cre and cardiotoxin (CTX) were injected into the calf muscles of 9, 14, or 28-day-old caALK2fl/- or caALK2fl/fl mice. HO was measured by planar radiography or microCT at 14-28 days post-injury. In 9-day-old caALK2fl/- or caALK2fl/fl mice, single injections of 109 PFU Adex-cre and 0.3 μg of CTX were sufficient to induce extensive HO within 14 days post-injury. In 28-day-old mice, the doses were increased to 5 x 109 PFU Adex-cre and 3.0 μg of CTX to achieve similar consistency, but at a slower rate versus younger mice. Using a crush injury, instead of CTX, also provided consistent induction of HO. Finally, the Type 1 BMPR inhibitor, DMH1, significantly reduced HO formation in 28-day-old caALK2fl/fl mice. These data illustrate multiple methods for reliable induction of localized HO in the caALK2flmouse that can serve as a starting point for new laboratories utilizing this model.
Sigma-Aldrich
DMH1, ≥98% (HPLC)
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin solution