MilliporeSigma
  • Tropomyosin isoforms regulate cofilin 1 activity by modulating actin filament conformation.
Tropomyosin isoforms regulate cofilin 1 activity by modulating actin filament conformation.
Archives of biochemistry and biophysics (2020-01-31)
Zofia Ostrowska-Podhorodecka, Małgorzata Śliwinska, Emil Reisler, Joanna Moraczewska
ABSTRACT
Tropomyosin and cofilin are involved in the regulation of actin filament dynamic polymerization and depolymerization. Binding of cofilin changes actin filaments structure, leading to their severing and depolymerization. Non-muscle tropomyosin isoforms were shown before to differentially regulate the activity of cofilin 1; products of TPM1 gene stabilized actin filaments, but products of TPM3 gene promoted cofilin-dependent severing and depolymerization. Here, conformational changes at the longitudinal and lateral interface between actin subunits resulting from tropomyosin and cofilin 1 binding were studied using skeletal actin and yeast wild type and mutant Q41C and S265C actins. Cross-linking of F-actin and fluorescence changes in F-actin labeled with acrylodan at Cys41 (in D-loop) or Cys265 (in H-loop) showed that tropomyosin isoforms differentially regulated cofilin-induced conformational rearrangements at longitudinal and lateral filament interfaces. Tryptic digestion of F-Mg-actin confirmed the differences between tropomyosin isoforms in their regulation of cofilin-dependent changes at actin-actin interfaces. Changes in the fluorescence of AEDANS attached to C-terminal Cys of actin, as well as FRET between Trp residues in actin subdomain 1 and AEDANS, did not show differences in the conformation of the C-terminal segment of F-actin in the presence of different tropomyosins ± cofilin 1. Therefore, actin's D- and H-loop are the sites involved in regulation of cofilin activity by tropomyosin isoforms.