MilliporeSigma
  • Immunoaffinity chromatography of abscisic acid combined with electrospray liquid chromatography-mass spectrometry.
Immunoaffinity chromatography of abscisic acid combined with electrospray liquid chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-10-27)
Veronika Hradecká, Ondrej Novák, Libor Havlícek, Miroslav Strnad
ABSTRACT
Polyclonal antibodies with high specificity for C1-immobilised (+)-cis,trans-abscisic acid (ABA) were raised, characterised by enzyme-linked immunosorbent assay (ELISA) and used for preparation of an immunoaffinity chromatography (IAC) gel. The detection limit of the ELISA was approximately 4.6x10(-10)mol/L. Sensitive electrospray liquid chromatography-mass spectrometry (LC-ESI-MS) methods were also developed with detection limits below 0.1x10(-12)mol. The IAC allowed quick, single-step processing of samples prior to the analyses. The LC-ESI-MS and LC-ELISA techniques were used for comparative estimation of endogenous ABA levels in immunoaffinity purified extracts of normal and water-stressed Nicotiana tabacum L. leaves. The analytical approaches were validated using deuterium- and tritium-labelled internal standards, respectively. The IAC method was found to be highly effective, sensitive and convenient for isolating the target analyte from plant material.