MilliporeSigma
  • Adenosine-5'-tetraphosphate and guanosine-5'-tetraphosphate: new substrates of the cytosolic exopolyphosphatase of the yeast Saccharomyces cerevisiae.
Adenosine-5'-tetraphosphate and guanosine-5'-tetraphosphate: new substrates of the cytosolic exopolyphosphatase of the yeast Saccharomyces cerevisiae.
Biochemistry. Biokhimiia (1998-02-11)
T V Kulakovskaya, N A Andreeva, I S Kulaev
ABSTRACT
A cytosolic preparation of Saccharomyces cerevisiae is capable of hydrolyzing adenosine-5'-tetraphosphate and guanosine-5'-tetraphosphate with activities which are 1.5-2 times greater than that with polyP15. The apparent K(m) values for hydrolysis of adenosine-5'-tetraphosphate and guanosine-5'-tetraphosphate are 100 and 80 microM, respectively. A comparative study of inhibitors shows that these activities are inherent characteristics of these exopolyphosphatases.