MilliporeSigma
  • Genome-wide methylation analysis of tubulocystic and papillary renal cell carcinomas.
Genome-wide methylation analysis of tubulocystic and papillary renal cell carcinomas.
Neoplasma (2016-03-02)
M Korabecna, J Geryk, M Hora, P Steiner, O Seda, V Tesar
ABSTRACT
Tubulocystic renal cell carcinoma (TRCC) represents a rare tumor with incidence lower than 1 % of all renal carcinomas. This study was undertaken to contribute to characterization of molecular signatures associated with TRCC and to compare them with the features of papillary renal cell carcinoma (PRCC) at the level of genome wide methylation analysis.We performed methylated DNA immunoprecipitation (MeDIP) coupled with microarray analysis (Roche NimbleGen). Using the CHARM package, we compared the levels of gene methylation between paired samples of tumors and control renal tissues of each examined individual. We found significant global demethylation in all tumor samples in comparison with adjacent kidney tissues of normal histological appearance but no significant differences in gene methylation between the both compared tumor entities. Therefore we focused on characterization of differentially methylated regions between both tumors and control tissues. We found 42 differentially methylated genes.Hypermethylated genes for protocadherins (PCDHG) and genes coding for products associated with functions of plasma membrane were evaluated as significantly overrepresented among hypermethylated genes detected in both types of renal cell carcinomas.In our pilot study, we provide the first evidence that identical features in the process of carcinogenesis leading to TRCC and/or to PRCC may be found at the gene methylation level.
Sigma-Aldrich
GenomePlex® Complete Whole Genome Amplification (WGA) Kit, Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue
Pricing and availability is not currently available.