MilliporeSigma
  • Quantifying phthalate metabolites in human meconium and semen using automated off-line solid-phase extraction coupled with on-line SPE and isotope-dilution high-performance liquid chromatography--tandem mass spectrometry.
Quantifying phthalate metabolites in human meconium and semen using automated off-line solid-phase extraction coupled with on-line SPE and isotope-dilution high-performance liquid chromatography--tandem mass spectrometry.
Analytical chemistry (2006-09-15)
Kayoko Kato, Manori J Silva, Larry L Needham, Antonia M Calafat
ABSTRACT
We developed an analytical method using off-line solid-phase extraction (SPE) coupled with on-line SPE and isotope-dilution high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) to determine the concentrations of phthalate metabolites in human meconium and in semen. First, we used off-line SPE to remove interfering proteins and other biomolecules from the samples. Then, we preconcentrated the phthalate metabolites in the extract using on-line SPE before measuring them by HPLC-MS/MS. For most of the analytes, the limits of detection ranged between 0.2 and 0.7 ng/g for meconium and between 0.3 and 0.7 ng/mL for semen. The recovery after off-line SPE varied for most analytes between 65 and 99% at concentrations ranging from 3.0 to 30.0 ng/mL in semen and between 67 and 103% at concentrations ranging from 2.0 to 10.0 ng/mL in meconium. Precision measured by the relative standard deviation ranged from 3.2 to 19.1% for intraday and from 3.9 to 18.6% for interday. We validated this novel approach--which is applicable to other biological matrixes, including serum and breast milk--on spiked samples and on five meconium samples and one pooled semen sample from people with no known occupational exposure to phthalates.
Sigma-Aldrich
Monoisobutyl phthalate, ≥95%
Pricing and availability is not currently available.