MilliporeSigma
  • Essential arginine residues in the active sites of propionyl CoA carboxylase and beta-methylcrotonyl CoA carboxylase.
Essential arginine residues in the active sites of propionyl CoA carboxylase and beta-methylcrotonyl CoA carboxylase.
Enzyme (1979-01-01)
B Wolf, F Kalousek, L E Rosenberg
PMID510274
ABSTRACT
At least one arginine residue is essential for substrate binding in or near the active sites of propionyl CoA carboxylase (PCC) and beta-methylcrotonyl CoA carboxylase (beta MCC) in cultured human fibroblasts. This conclusion is based on studies of enzyme inhibition by phenylglyoxal, a reagent which specifically modifies arginine residues. Human fibroblast PCC both in extracts and in a 20-fold purified preparation was nearly completely protected from phenylglyoxal inhibition following incubation with propionyl CoA or ATP. It appears that a phosphate group from either ATP or the CoA moiety of propionyl CoA reacts with the essential arginine residue(s). beta MCC which was similarly inhibited by phenylglyoxal was protected by beta-methylcrotonyl CoA and ATP. Thus phenylglyoxal may be used to label specific arginine residues within the active sites of previously sequenced carboxylases.
Sigma-Aldrich
β-Methylcrotonyl coenzyme A lithium salt, ≥90%
Pricing and availability is not currently available.