MilliporeSigma
  • Updated occurrence of 3-monochloropropane-1,2-diol esters (3-MCPD) and glycidyl esters in infant formulas purchased in the United States between 2017 and 2019.
Updated occurrence of 3-monochloropropane-1,2-diol esters (3-MCPD) and glycidyl esters in infant formulas purchased in the United States between 2017 and 2019.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2020-01-10)
Jessica K Beekman, Kaitlin Grassi, Shaun MacMahon
ABSTRACT
Fatty acid esters of 3-monochloropropane-1,2-diol (3-MCPD) and glycidol are potentially carcinogenic and/or genotoxic processing contaminants that are formed during the process of edible oil refining. Because of their toxicological properties, the presence of these compounds in refined oils and foods containing these oils, particularly infant formula, poses a potential food safety concern. For this reason, recent research efforts have focussed on the development of methods for the analysis of MCPD and glycidyl esters in infant formula in order to estimate levels of exposure. This work presents occurrence data for 3-MCPD and glycidyl esters in 222 infant formulas purchased in the United States between December 2017 and January 2019. The results of this study show a wide range of contaminant concentrations across four different manufacturers, with average bound 3-MCPD concentrations ranging from 0.035 µg g-1 to 0.63 µg g-1 and average bound glycidol concentrations ranging from 0.019 µg g-1 to 0.22 µg g-1. The data suggest that manufacturers B and C source palm oil produced with mitigation measures, leading to reduced amounts of 3-MCPD and glycidyl esters in their infant formulas. Additionally, comparison with a previously published study in our laboratory of the occurrence of 3-MCPD and glycidyl esters in infant formula purchased in the U.S. between 2013 and 2016 revealed that, since 2016, contaminant concentrations have decreased in products produced by manufacturers A, B, and C, while contaminant amounts in formulas from manufacturer D have slightly increased.