MilliporeSigma
  • Effects of Dietary Fiber Sources during Gestation on Stress Status, Abnormal Behaviors and Reproductive Performance of Sows.
Effects of Dietary Fiber Sources during Gestation on Stress Status, Abnormal Behaviors and Reproductive Performance of Sows.
Animals : an open access journal from MDPI (2020-01-19)
Shuangbo Huang, Jianfu Wei, Haoyuan Yu, Xiangyu Hao, Jianjun Zuo, Chengquan Tan, Jinping Deng
ABSTRACT
Inclusion of fiber in gestation diets is a method for enhancing satiety and reducing abnormal behaviors in restricted feeding sows without providing excess energy. The purpose of this study was to use an in vitro-in vivo method to appraise the effects of two available unconventional dietary fiber resources during gestation on sows' physio-chemical properties of diets, postprandial satiety, performance, abnormal behaviors, stress status and lactation feed intake under three different dietary treatments: control diet (CON diet), 5% resistant starch diet (RS diet), and 5% fermented soybean fiber diet (FSF diet) with a total of 78 (average parity 5) Landrace × Yorkshire sows. Results showed that swelling capacity was higher in the RS diet than in the CON or FSF diet. Meanwhile, the 48 h cumulative gas production and the final asymptotic gas volume after in vitro fermentation of gestation diets showed an increased trend (p = 0.07, p = 0.09, respectively) in the RS diet versus the CON or FSF diets. While the sows' litter size, body weight, backfat or weaning-to-estrus interval were not affected (p > 0.05) by the three treatments during gestation, the RS group showed a decline in stillbirth number (p < 0.05) and stillbirth rate (p < 0.01) relative to the other two groups. Meanwhile, the proportion of standing was lower while the sow's serum concentrations of PYY (peptide YY) and GLP-1 (glucagon-like peptide-1) were higher (p < 0.05) on day 70 of gestation in the RS group than in the CON or FSF group. Compared with the CON group, the RS group showed a downward tendency (p = 0.07) in the sows' plasma cortisol concentration on day 70 of gestation. A comparison of oxidative and antioxidative indicators revealed an increase in the sows' serum FRAP (ferric ion reducing antioxidant power) (p < 0.05) and a decrease of protein carbonyl (p < 0.05) on day 109 of gestation in the RS or FSF group versus the CON group. Overall, inclusion of 5% RS with greater swelling capacity in the gestation diet contributed to enhancing the postprandial satiety, alleviating the stress status, reducing the abnormal behaviors and thus lowering the stillbirth rate of sows.