MilliporeSigma
  • A Novel Tissue and Stem Cell Specific TERF1 Splice Variant Is Downregulated in Tumour Cells.
A Novel Tissue and Stem Cell Specific TERF1 Splice Variant Is Downregulated in Tumour Cells.
International journal of molecular sciences (2019-12-28)
Yousef Ashraf Tawfik Morcos, Gregoire Najjar, Sabine Meessen, Britta Witt, Anca Azoitei, Mukesh Kumar, Gamal Wakileh, Klaus Schwarz, Hubert Schrezenmeier, Friedemann Zengerling, Christian Bolenz, Cagatay Günes
ABSTRACT
In this study, we describe the identification of a novel splice variant of TERF1/PIN2, one of the main components of the telomeric shelterin complex. This new splice variant is identical to TERF1, apart from a 30 amino acid internal insertion near to the C-terminus of TERF1. Based on genome comparison analyses and RNA expression data, we show that this splice variant is conserved among hominidae but absent from all other species. RNA expression and histological analyses show specific expression in human spermatogonial and hematopoietic stem cells (HSCs), while all other analyzed tissues lack the expression of this TERF1-isoform, hence the name TERF1-tsi (TERF1-tissue-specific-isoform). In addition, we could not detect any expression in primary human cells and established cancer cell lines. Immunohistochemistry results involving two new rabbit polyclonal antibodies, generated against TERF1-tsi specific peptides, indicate nuclear localization of TERF1-tsi in a subset of spermatogonial stem cells. In line with this observation, immunofluorescence analyzes in various cell lines consistently revealed that ectopic TERF1-tsi localizes to the cell nucleus, mainly but not exclusively at telomeres. In a first attempt to evaluate the impact of TERF1-tsi in the testis, we have tested its expression in normal testis samples versus matched tumor samples from the same patients. Both RT-PCR and IHC show a specific downregulation of TERF1-tsi in tumor samples while the expression of TERF1 and PIN2 remains unchanged.