MilliporeSigma
  • PAMAM-cRGD mediating efficient siRNA delivery to spermatogonial stem cells.
PAMAM-cRGD mediating efficient siRNA delivery to spermatogonial stem cells.
Stem cell research & therapy (2019-12-20)
Tianjiao Li, Qiwen Chen, Yi Zheng, Pengfei Zhang, Xiaoxu Chen, Junna Lu, Yinghua Lv, Shiguo Sun, Wenxian Zeng
ABSTRACT
Spermatogonial stem cells (SSCs) are the cornerstone of sperm production and thus perpetual male fertility. In clinics, transplantation of patient's own SSCs into testes is a promising technique to restore fertility when male germ cells have been depleted by gonadotoxic therapies. Auto-transplantation of genetically modified SSCs even has the potential to treat male infertility caused by genetic mutations. However, SSCs are refractory to transfection approaches. Poly(amidoamine) (PAMAM) dendrimers have the unique three-dimensional architecture, surface charge, and high density of surface groups that are suitable for ligand attachment, thereby facilitating target delivery. The goal of this study was to elucidate whether PAMAM dendrimers can efficiently deliver short interfering RNAs (siRNAs) to SSCs. We introduced cyclic arginine-glycine-aspartic acid (cRGD) peptides to the fifth generation of PAMAM dendrimers (G5) to generate PAMAM-cRGD dendrimers (G5-cRGD). The characterization of G5-cRGD was detected by Fourier transform infrared spectroscope (FTIR), transmission electron microscope (TEM), and the Cell Counting Kit-8 (CCK-8) assay. Confocal microscopy and flow cytometry were used to evaluate the delivery efficiency of siRNA by G5-cRGD to SSCs. The results showed that G5-cRGD encompassing siRNA could self-assemble into spherical structures with nanoscale size and possess high transfection efficiency, excellent endosomal escape ability, and low cytotoxicity, superior to a commercial transfection reagent Lipofectamine® 2000. Moreover, we demonstrated that G5-cRGD efficiently delivered siRNAs and triggered gene silencing. This study thus provides a promising nanovector for siRNA delivery in SSCs, facilitating the future clinical application of SSC auto-transplantation with genetically modified cells with a hope to cure male infertility that is caused by genetic disorders.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Donkey Anti-Mouse IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Pricing and availability is not currently available.