MilliporeSigma
  • Apoptosis induction by T-2 toxin: activation of caspase-9, caspase-3, and DFF-40/CAD through cytosolic release of cytochrome c in HL-60 cells.
Apoptosis induction by T-2 toxin: activation of caspase-9, caspase-3, and DFF-40/CAD through cytosolic release of cytochrome c in HL-60 cells.
Bioscience, biotechnology, and biochemistry (2001-10-02)
M Nagase, M M Alam, A Tsushima, T Yoshizawa, N Sakato
ABSTRACT
The molecules participating in apoptosis induced by T-2 toxin in human leukemia HL-60 cells were investigated. The rank order of the potency of trichothecene mycotoxins to induce internucleosomal DNA fragmentation was found to be T-2, satratoxin G, roridin A > diacetoxyscirpenol > baccharin B-5 > nivalenol, deoxynivalenol, 3-acetyldeoxynivalenol, fusarenon-X, baccharin B-4=vehicle control. Western blot analysis of caspase-3 in T-2-treated cells clearly indicated the appearance of its catalytically active fragment of 17-kDa. Increased caspase-3 activity was also detected by using a fluorogenic substrate, DEVD-AMC. Next, cells exposed to T-2 led to cleavage of PARP from its native 116-kDa form to the 85-kDa product. Moreover, DFF-45/ICAD were cleaved to give a 12.5-kDa fragment via T-2 treatment. T-2 caused the release of cytochrome c from mitochondria into the cytosol. Increased enzymic activity of caspase-9 on LEHD-AMC was shown. These data indicate that T-2-induced apoptosis involves activation of caspase-3 and DFF-40/CAD through cytosolic accumulation of cytochrome c along with caspase-9 activation.