MilliporeSigma
  • Position 2 and position 2/Ala15-substituted analogs of bovine growth hormone-releasing factor (bGRF) with enhanced metabolic stability and improved in vivo bioactivity.
Position 2 and position 2/Ala15-substituted analogs of bovine growth hormone-releasing factor (bGRF) with enhanced metabolic stability and improved in vivo bioactivity.
Journal of medicinal chemistry (1993-04-02)
T M Kubiak, A R Friedman, R A Martin, A K Ichhpurani, G R Alaniz, W H Claflin, M C Goodwin, D L Cleary, C R Kelly, R M Hillman
ABSTRACT
In order to prepare GRF analogs with high activity in vivo, a strategy was undertaken to stabilize the peptide to dipeptidylpeptidase IV (DPP-IV), a protease found in plasma which inactivates native human and bovine GRF by cleavage of the Ala2-Asp3 bond. Replacement of the Ala2 residue with Ser, Thr, or Gly in [Leu27]bGRF(1-29)NH2 resulted in peptides greatly stabilized against proteolysis in plasma, but having low inherent GH-releasing activity when tested in bovine pituitary cell cultures. Replacement of Gly15 with Ala15 was marginally effective in improving the in vitro bioactivity of this group of peptides. When tested for GH-hormone release in steers, however, the Thr2,Ala15 analog was four times more potent than bGRF(1-44)NH2. Eleven additional analogs from the [X2,Ala15,Leu27]bGRF(1-29)NH2 series were synthesized and evaluated for metabolic stability in bovine plasma and for GH releasing activity in steers in vivo and in rat pituitary cells in vitro. Two compounds, [Val2,Ala15,Leu27]dGRF(1-29)NH2 and [Ile2,Ala15,Leu27]-bGRF(1-29)NH2, had increased GH-releasing activity in steers over that of [Thr2,Ala15,Leu27]-bGRF(1-29)NH2 and over a previously reported super-potent analog, [desNH2Tyr1,D-Ala2,Ala15]-hGRF(1-29)NH2.
Sigma-Aldrich
[His1,Nle27]-Growth Hormone Releasing Factor 1-32 amide human
Sigma-Aldrich
[D-Ala2]-Growth Hormone Releasing Factor 1-29 amide human