Merck
  • Direct-Write Bioprinting Approach to Construct Multilayer Cellular Tissues.
Direct-Write Bioprinting Approach to Construct Multilayer Cellular Tissues.
Frontiers in bioengineering and biotechnology (2020-02-11)
Elahe Masaeli, Christophe Marquette
ABSTRACT
As a cellular-assembly technique, bioprinting has been extensively used in tissue engineering and regenerative medicine to construct hydrogel-based three-dimensional (3D) tissue-like models with prescribed geometry. Here, we introduced a unique direct-write bioprinting strategy to fabricate a bilayer flat tissue in a hydrogel-free approach. A printed retina pigmented epithelium layer (RPE) was applied as living biopaper for positioning a fibroblast layer without using any hydrogel in bioink. We adjusted the number of cells in the inkjet droplets in order to obtain a uniform printed cell layer and demonstrated the formation of a bilayer construct through confocal imaging. Since our printing system introduced low levels of shear stress to the cells, it did not have a negative effect on cell survival, although cell viability was generally lower than that of control group over 1 week post-printing. In conclusion, our novel direct-write bioprinting approach to spatiotemporally position different cellular layers may represent an efficient tool to develop living constructs especially for regeneration of complex flat tissues.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Resazurin based