MilliporeSigma
  • miR-1303 promotes the proliferation, migration and invasion of prostate cancer cells through regulating the Wnt/β-catenin pathway by targeting DKK3.
miR-1303 promotes the proliferation, migration and invasion of prostate cancer cells through regulating the Wnt/β-catenin pathway by targeting DKK3.
Experimental and therapeutic medicine (2019-11-28)
Bo Liu, Weidong Zhou, Huiyang Jiang, Zhendong Xiang, Lei Wang
ABSTRACT
MicroRNA-1303 (miR-1303) is involved in the tumorigenesis and progression of several cancers, and yet the role of miR-1303 in prostate cancer (PCa) and its underlying mechanism are unknown. To explore this issue, the present study aimed to use PCa tissues, cell lines and a PCa-engrafted mouse model to determine the expression and roles of miR-1303 in PCa. Furthermore, a series of experiments were conducted to explore the underlying mechanisms of action of miR-1303 in PCa cells. miR-1303 was demonstrated to be highly expressed in PCa tissues and cell lines. The level of miR-1303 expression was closely associated with higher Gleason scores and a more developed tumor stage in patients with PCa, and patients with higher levels of miR-1303 displayed a reduced overall survival rate. miR-1303 overexpression promoted the proliferation, migration and invasion of PCa cells. In vivo experiments showed that miR-1303 inhibition suppressed the growth of PCa tumors in mice. Additionally, dickkopf Wnt signaling pathway inhibitor 3 (DKK3) was identified as a target of miR-1303. Knockdown of miR-1303 suppressed the proliferation, migration and invasion of PCa cells, increased DKK3 expression, and inhibited the activity of the Wnt/β-catenin pathway. In conclusion, miR-1303 may promote proliferation, migration and invasion of PCa cells through activating the Wnt/β-catenin pathway by regulating DKK3 expression. These results indicated that miR-1303 may be considered as a potential biomarker for PCa treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human DKK3