MilliporeSigma
  • Increased IL-26 Expression Promotes T Helper Type 17- and T Helper Type 2-Associated Cytokine Production by Keratinocytes in Atopic Dermatitis.
Increased IL-26 Expression Promotes T Helper Type 17- and T Helper Type 2-Associated Cytokine Production by Keratinocytes in Atopic Dermatitis.
The Journal of investigative dermatology (2019-08-30)
Hiroaki Kamijo, Tomomitsu Miyagaki, Yoshio Hayashi, Taro Akatsuka, Sayaka Watanabe-Otobe, Tomonori Oka, Naomi Shishido-Takahashi, Hiraku Suga, Makoto Sugaya, Shinichi Sato
ABSTRACT
Whereas atopic dermatitis (AD) is considered as a T helper 2 (Th2)-centered disease, IL-17-producing Th (Th17) cells are also activated in AD lesional skin. However, the relationship between Th17 responses and Th2 responses in AD is still to be elucidated. Although Th17 cells are increased in AD skin, the expression and function of IL-26, which is also produced by Th17 cells, in AD are still unknown. In this report, we demonstrated that IL-26 mRNA expression levels were elevated in AD lesional skin compared with healthy controls and that IL-26-producing cells were increased in AD lesional skin by immunohistochemistry. Furthermore, IL-26 promoted IL-8, IL-1β, chemokine (C-C motif) ligand 20, IL-33, and β-defensin 2 production in keratinocytes through phosphorylation of signal transducer and activator of transcription 1 and signal transducer and activator of transcription 3. Selective JAK inhibitors for JAK1, JAK2, and tyrosine kinase 2 blocked IL-26-induced cytokine production in keratinocytes. We also showed that injection of IL-26 exacerbated an oxazolone-induced AD mouse model and upregulated Th2 and Th17 cytokine expression in vivo. Because previous studies indicate that the above molecules induced by IL-26 can promote Th17 and/or Th2 immune responses, IL-26 may play an important role for bridging between Th17 and Th2 responses, resulting in the development of AD.
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)