MilliporeSigma
  • Discrete Supertetrahedral T5 Selenide Clusters and Their Se/S Solid Solutions: Ionic-Liquid-Assisted Precursor Route Syntheses and Photocatalytic Properties.
Discrete Supertetrahedral T5 Selenide Clusters and Their Se/S Solid Solutions: Ionic-Liquid-Assisted Precursor Route Syntheses and Photocatalytic Properties.
Chemistry (Weinheim an der Bergstrasse, Germany) (2020-01-24)
Yanqi Wang, Zhipeng Zhu, Zhaofeng Sun, Qianqian Hu, Jianrong Li, Jiang Jiang, Xiaoying Huang
ABSTRACT
Although supertetrahedral Tn sulfide clusters (n=2-6) have been extensively explored, the synthesis of Tn selenide clusters with n>4 has not been achieved thus far. Reported here are ionic-liquid (IL)-assisted precursor route syntheses, characterizations, and the photocatalytic properties of six new M-In-Q (M=Cu or Cd; Q=Se or Se/S) chalcogenide compounds, namely [Bmmim]12 Cu5 In30 Q52 Cl3 (Im) (Q=Se (T5-1), Se48.5 S3.5 (T5-2); Bmmim=1-butyl-2,3-dimethylimidazolium, Im=imidazole), [Bmmim]11 Cd6 In28 Q52 Cl3 (MIm) (Q=Se (T5-3), Se28.5 S23.5 (T5-4), Se16 S36 (T5-5); MIm=1-methylimidazole), and [Bmmim]9 Cd6 In28 Se8 S44 Cl(MIm)3 (T5-6). The cluster compounds T5-1 and T5-3 represent the largest molecular supertetrahedral Tn selenide clusters to date. Under visible-light illumination, the Cu-In-Q compounds showed photocatalytic activity towards the decomposition of crystal violet, whereas the Cd-In-Q compounds exhibited good photocatalytic H2 evolution activity. Interestingly, the experimental results show that the photocatalytic performances of the selenide/sulfide solid solutions were significantly better than those of their selenide analogues, for example, the degradation time of the organic dye with T5-2 was much shorter than that with T5-1, whereas the photocatalytic H2 evolution efficiencies with T5-3-T5-6 improved significantly with increasing sulfur content. This work highlights the significance of IL-assisted precursor route synthesis and the tuning of photocatalytic properties through the formation of solid solutions.
Sigma-Aldrich
1-Butyl-2,3-dimethylimidazolium chloride, ≥97.0% (HPLC/AT)
Pricing and availability is not currently available.