MilliporeSigma
  • Downregulation of aryl hydrocarbon receptor expression decreases gastric cancer cell growth and invasion.
Downregulation of aryl hydrocarbon receptor expression decreases gastric cancer cell growth and invasion.
Oncology reports (2013-04-23)
Xiao-Fei Yin, Jie Chen, Wei Mao, Yu-Hong Wang, Min-Hu Chen
ABSTRACT
Aryl hydrocarbon receptor (AhR) is a ligand-activated transcription factor associated with tumor initiation and progression. AhR expression is significantly increased in gastric cancer tissues and gastric cancer cell lines; however, the relationship between AhR and gastric cancer is still unclear. In the present study, we explored the effects of the inhibition of AhR expression by RNA interference on the biological behavior of gastric cancer cells (MKN45 and SGC7901), and elucidated the specific mechanisms of AhR action in the development of gastric cancer. Results showed that small interfering RNA (siRNA) against AhR effectively inhibited the expression of AhR, and decreased the expression of cytochrome P450 (CYP)1A1 and CYP1B1, which are classic target genes of the AhR pathway. Compared to the negative control group, AhR-siRNA-transfected cells showed decreased cellular growth, delayed G1-S cell cycle progression and increased apoptosis rate. Furthermore, inhibition of AhR expression by siRNA in SGC7901 cells led to decreased cell migratory and invasive ability, accompanied by downregulation of expression and activity of matrix metalloproteinase (MMP)-2 and MMP-9. Our results, therefore, suggest that AhR promotes the growth and invasiveness of gastric cancer cells and AhR may serve as a promising therapeutic target for gastric cancer.
Sigma-Aldrich
Anti-Cytochrome P450 Enzyme CYP1A1 Antibody, serum, Chemicon®
Pricing and availability is not currently available.