MilliporeSigma
  • Study of the Royal Jelly Free Fatty Acids by Liquid Chromatography-High Resolution Mass Spectrometry (LC-HRMS).
Study of the Royal Jelly Free Fatty Acids by Liquid Chromatography-High Resolution Mass Spectrometry (LC-HRMS).
Metabolites (2020-01-23)
Maroula G Kokotou, Christiana Mantzourani, Rodalia Babaiti, George Kokotos
ABSTRACT
The lipidome of royal jelly (RJ) consists of medium-chained (8-12 carbon atoms) free fatty acids. We present herein a liquid chromatography-high resolution mass spectrometry (HRMS) method that permits the determination of RJ fatty acids and at the same time the detection of suspect fatty acids. The method allows for the direct quantification of seven free fatty acids of RJ, avoiding any derivatization step. It was validated and applied in seven RJ samples, where the major RJ fatty acid trans-10-hydroxy-2-decenoic acid (10-HDA) was found to vary from 0.771 ± 0.08 to 0.928 ± 0.04 g/100 g fresh RJ. Four additional suspect fatty acids were simultaneously detected taking advantage of the HRMS detection.
Sigma-Aldrich
(±)-3-Hydroxydecanoic acid, ≥98%