MilliporeSigma
  • A new highly transparent injectable PHA-based thermogelling vitreous substitute.
A new highly transparent injectable PHA-based thermogelling vitreous substitute.
Biomaterials science (2019-12-14)
Kun Xue, Zengping Liu, Lu Jiang, Dan Kai, Zibiao Li, Xinyi Su, Xian Jun Loh
ABSTRACT
Transparency is an important criterion for the application of biomaterials to the eye and essential for use as a vitreous substitute. However, there is a lack of understanding on the features of transparent hydrogels and the boundaries of transparency of various hydrogel systems. In this paper, we tune the poly[(R)-3-hydroxybutyrate-(R)-3-hydroxyhexanoate] (PHBHx) content of biodegradable PHBHx-based polyurethane thermogels and show that the amount of hydrophobic PHBHx correlates with hydrogel cloudiness. We found that PHxEP-0.5 hydrogel shows high light transmittance and suitable gel properties as a vitreous substitute. The PHxEP-0.5 hydrogel is able to maintain transparency when implanted in rabbit eyes as opposed to a cloudy gel and shows negligible inflammation and preservation of the retinal structure over 6 months. In conclusion, we show that PHBHx-based thermogels can be tuned for transparency and are biocompatible in the rabbit eye. These results could be instructive for the design of a new generation of injectable transparent vitreous substitutes.
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%