MilliporeSigma
  • The role of the DNA damage checkpoint pathway in intraductal papillary mucinous neoplasms of the pancreas.
The role of the DNA damage checkpoint pathway in intraductal papillary mucinous neoplasms of the pancreas.
Clinical cancer research : an official journal of the American Association for Cancer Research (2007-08-03)
Yoshihiro Miyasaka, Eishi Nagai, Hiroshi Yamaguchi, Kei Fujii, Takahiro Inoue, Kenoki Ohuchida, Tomomi Yamada, Kazuhiro Mizumoto, Masao Tanaka, Masazumi Tsuneyoshi
ABSTRACT
Intraductal papillary mucinous neoplasms (IPMN) are known to show a transition from adenoma to carcinoma accompanied by several molecular abnormalities. ATM-Chk2-p53 DNA damage checkpoint activation, which is involved in prevention of the progression of several tumors, was analyzed to evaluate the role of the DNA damage checkpoint in the progression of IPMNs. One hundred and twenty-eight IPMNs were classified into four groups (intraductal papillary mucinous adenoma, borderline IPMN, noninvasive intraductal papillary mucinous carcinoma, and invasive intraductal papillary mucinous carcinoma) and stained immunohistochemically using antibody for Thr(68)-phosphorylated Chk2. Expression of ATM, Chk2, and p21(WAF1) and accumulation of p53 were also analyzed. Chk2 phosphorylation was shown in all adenomas and showed a significant decreasing trend with the progression of atypia (P < 0.0001 by the Cochran-Armitage test for trend). Expression of p21(WAF1) also exhibited a decreasing tendency (P < 0.0001), reflecting DNA damage checkpoint inactivation. p53 accumulation was mostly detected in malignant IPMNs. It was suggested that the DNA damage checkpoint provides a selective pressure for p53 mutation. Our findings indicate that DNA damage checkpoint activation occurs in the early stage of IPMNs and prevents their progression. It is suggested that disturbance of the DNA damage checkpoint pathway due to Chk2 inactivation or p53 mutation contributes to the carcinogenesis of IPMNs.
Sigma-Aldrich
Anti-p21WAF1 Antibody, clone EA10, clone EA10, from mouse