MilliporeSigma
  • The E3 SUMO ligase PIASy is a regulator of cellular senescence and apoptosis.
The E3 SUMO ligase PIASy is a regulator of cellular senescence and apoptosis.
Molecular cell (2006-06-24)
Oliver Bischof, Klaus Schwamborn, Nadine Martin, Andreas Werner, Claudio Sustmann, Rudolf Grosschedl, Anne Dejean
ABSTRACT
Cellular senescence and apoptosis have evolved to restrain unwarranted proliferation of potentially tumorigenic cells. Here we show that overexpression of the E3 SUMO ligase PIASy in normal human fibroblasts recruits the p53 and Rb tumor suppressor pathways to provoke a senescence arrest. By contrast, in Rb-deficient fibroblasts, expression of PIASy leads to p53-dependent apoptosis. Induction of senescence requires PIASy E3 activity and is specific for this member of the PIAS ligase family. PIASy stimulates sumoylation and transcriptional activity of p53 and increases Rb-dependent corepression through recruitment to E2F-responsive promoters. Viral oncoprotein E6 suppresses both PIASy-induced senescence and sumoylation of PIASy substrates. Finally, we show that fibroblasts lacking PIASy exhibit a highly reduced propensity to undergo senescence in response to a prosenescence stimulus. Altogether, these data provide the first evidence for a direct role of an E3 SUMO ligase, and by implication of the SUMO pathway, in cellular senescence and apoptosis.
Sigma-Aldrich
Anti-MDM2 Antibody, clone 2A10, clone 2A10, from mouse
Pricing and availability is not currently available.