MilliporeSigma
  • DNA-mediated adjuvant immunotherapy extends survival in two different mouse models of myeloid malignancies.
DNA-mediated adjuvant immunotherapy extends survival in two different mouse models of myeloid malignancies.
Oncotarget (2015-09-18)
Carole Le Pogam, Satyananda Patel, Petra Gorombei, Laura Guerenne, Patricia Krief, Nader Omidvar, Nilgun Tekin, Elena Bernasconi, Flore Sicre, Marie-Helene Schlageter, Martine Chopin, Maria-Elena Noguera, Robert West, Ansu Abu, Vikram Mathews, Marika Pla, Pierre Fenaux, Christine Chomienne, Rose Ann Padua
ABSTRACT
We have previously shown that a specific promyelocytic leukemia-retinoic acid receptor alpha (PML-RARA) DNA vaccine combined with all-trans retinoic acid (ATRA) increases the number of long term survivors with enhanced immune responses in a mouse model of acute promyelocytic leukemia (APL). This study reports the efficacy of a non-specific DNA vaccine, pVAX14Flipper (pVAX14), in both APL and high risk myelodysplastic syndrome (HR-MDS) models. PVAX14 is comprised of novel immunogenic DNA sequences inserted into the pVAX1 therapeutic plasmid. APL mice treated with pVAX14 combined with ATRA had increased survival comparable to that obtained with a specific PML-RARA vaccine. Moreover, the survival advantage correlated with decreased PML-RARA transcript levels and increase in anti-RARA antibody production. In HR-MDS mice, pVAX14 significantly improved survival and reduced biomarkers of leukemic transformation such as phosphorylated mitogen-activated protein/extracellular signal-regulated kinase kinase (MEK) 1. In both preclinical models, pVAX14 vaccine significantly increased interferon gamma (IFNγ) production, memory T-cells (memT), reduced the number of colony forming units (CFU) and increased expression of the adapter molecule signalling to NF-κB, MyD88. These results demonstrate the adjuvant properties of pVAX14 providing thus new approaches to improve clinical outcome in two different models of myeloid malignancies, which may have potential for a broader applicability in other cancers.
Sigma-Aldrich
Anti-MEK1 Antibody, Upstate®, from rabbit